Anti-CD62L Rat Monoclonal Antibody (PerCP-Cyanine5.5) [clone: MEL-14]

Supplier: Tonbo Biosciences
65-0621-U025 65-0621-U100
75834-294EA 110 USD
75834-294 75834-296
Anti-CD62L Rat Monoclonal Antibody (PerCP-Cyanine5.5) [clone: MEL-14]
Antibodies
The MEL-14 antibody is specific for mouse CD62L, also known as L-Selectin, a cell adhesion molecule which facilitates lymphocyte "rolling" on activated vascular endothelium and homing to high endothelial venules (HEV) as immune cells transmigrate from blood into peripheral tissues. L-Selectin is a member of a family of Selectin molecules which act together with the integrin family of adhesion molecules to mediate leukocyte-endothelial interactions. L-Selectin is characteristically expressed by neutrophils, and is also found on B cells, monocytes, granulocytes, and at varying levels on naive, effector and memory T cells. It is rapidly shed upon cell activation, releasing into the circulation a soluble form whose biological role is of particular interest in cancer biology research. The MEL-14 antibody may be used as a phenotypic marker for CD62L expression on a variety of immune cell types. Please note that CD62L (L-Selectin) itself is also referred to as MEL-14 in the literature.

Type: Primary
Antigen: CD62L (L-Selectin)
Clonality: Monoclonal
Clone: MEL-14
Conjugation: PerCP-Cyanine5.5
Epitope:
Host: Rat
Isotype: Rat IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR