Anti-CD56 Mouse Monoclonal Antibody (FITC) [clone: MY31]

Supplier: Tonbo Biosciences
35-0564-T100 35-0564-T025
75834-270EA 205 USD
75834-270 75834-378
Anti-CD56 Mouse Monoclonal Antibody (FITC) [clone: MY31]
Antibodies
The MY31 antibody reacts with human CD56, also known as the neural cell adhesion molecule (NCAM), a glycoprotein which is a member of the immunoglobulin superfamily. The 140 kDa isoform of CD56 is expressed on human NK cells and NK-T cells, with increased expression levels on activated NK lymphocytes. The CD56 antigen is also expressed by neurons and is reported to play a role in nervous system development and neural cell-to-cell adhesion. Clone MY31 also reacts with a subset of CD14+ monocytes in non-human primates, and is reported to be cross-reactive with Chimpanzee, Cynomolgus and Rhesus.

Type: Primary
Antigen: CD56
Clonality: Monoclonal
Clone: MY31
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR