Nickel(II) cyanide tetrahydrate

Supplier: Pfaltz & Bauer
Danger

N04745-10G
101178-488EA 898.74 USD
101178-488
Nickel(II) cyanide tetrahydrate
Nickel(II) cyanide tetrahydrate
Formula: Ni(CN)₂·4H₂O
MW: 182.73 g/mol (110.73 anhy)
Boiling Pt: 100 °C
Melting Pt: 200 °C
MDL Number: MFCD00049490
CAS Number: 13477-95-7
UN: 1653
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR