Trithiocyanuric acid on resin (MP), fine beads, macroporous 100-200 mesh Loading: 1.7-1.8 mmol/g
Supplier: Supra Sciences
Trimercaptotriazine resin (TMT resin), highly effective scavenger of organometallic species.
Trimercaptotriazine Resin, MP, is a macroporous polystyrene resin functionalized with a trimercaptotriazine end group while its Silica counterpart is bound to a silica backbone. This reagent has shown to be an exceptional scavenger of organometallic reagents. MP-TMT is also extremely efficient in its ability to scavenge Copper. The limitations of solution phase trimercaptotriazine are based primarily around solubility in various solvents. The macroporous backbone of MP-TMT allows for its use in all solvents, and the Silica counterpart is effective in all solvents except any protic media. Simple filtration removes it (and the complexed metal) from your reaction.
Formula:
C₃H₃N₃S₃ MW: 177.27 g/mol |
MDL Number:
MFCD00006052 CAS Number: 638-16-4 |
Specification Test Results
Colour | Off-white |
Form | Granule |
Loading | 1.7-1.8 mmol/g |
Particle size | 100-200 mesh |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...