Water, Ultrapure cell & molecular grade, sterile
Supplier: Cytiva
Synonyms:
Dihydrogen oxide
WPW-CMG-1X6
WPW-CMG-1X6
75832-010CS
116.14
USD
CA75832-010
75832-010
Water, Ultrapure cell & molecular grade, sterile
Water
A universal, ultra pure water for your most sensitive research applications.
- This product is sterile and extremely low in endotoxin
- It is suitable for cell culture applications, media and reagent grade preparations
- Purified utilizing a nine step process resulting in very high quality water
This water has been analyzed for the absence of nucleases and proteases for your molecular biology applications. It is suitable for research investigations of nucleic acid and protein systems.
Certifications: Manufactured under cGMP and ISO 13485 guidelines in an FDA registered facility.
Formula:
H₂O MW: 18.02 g/mol |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 |
Specification Test Results
Appearance | Clear Liquid (Visual) |
Sterility |
Sterility (USP |
Endotoxin |
|
TOC (before packaging) |
|
Conductivity |
|
DNAse | None Detected |
RNAse | None Detected |
Protease | None Detected |
GMP manufactured | conform |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...