Water, certified for molecular biology
Supplier: Cytiva
Synonyms:
Dihydrogen oxide
WFI-MGW-20L
WFI-MGW-20L
10837-214EA
278
USD
CA10837-214
10837-214
Water, certified for molecular biology
Water
Cytiva Molecular Grade water is quality tested and certified RNase, DNase, and protease free.
- Purified utilizing a nine step process resulting in very high quality water
Every lot sent to our customer contains zero traces of toxic agents such as DEPC, and is well suited for use in diverse molecular biology applications.
Certifications: Manufactured under cGMP and ISO 13485 guidelines in an FDA registered facility.
Formula:
H₂O MW: 18.02 g/mol |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 |
Specification Test Results
Appearance | Clear Liquid (Visual) |
TOC |
|
Conductivity |
|
DNAse | None Detected |
Protease | None Detected |
RNAse | None Detected |
GMP manufactured | conform |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...