Water USP, Ph. Eur. for injection (WFI), sterile

Supplier: Cytiva

Synonyms: Dihydrogen oxide

WFI-EPZ-20L WFI-EPZ-2XL Z0104
10837-186EA 500.87 USD
10837-186 10837-188 75881-006
Water USP, Ph. Eur. for injection (WFI), sterile
Water

Cytiva's Water for Injection (WFI), multi-compendial grade is a terminally distilled, sterile, non-pyrogenic preparation which contains no bacteriostat, antimicrobial agent or added buffer. It complies with both USP and EP monographs for water for injection packaged in bulk for commercial use.


Manufactured under cGMP and ISO 13485 guidelines in an FDA registered facility.


Cytiva water is an excellent choice for upstream and downstream media/buffer preparation in biopharmaceutical manufacturing applications where EP requirements are necessary. It is also widely used in medical device manufacturing/processing applications where a high quality, sterile water is required.

Formula: H₂O
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR