National 210 Series Compound Microscopes
Supplier: National Optical
212-RLED 211-RLED
470213-088EA 638 USD
470213-088 470213-090
National 210 Series Compound Microscopes
Microscopes
Great Quality at a Great Price.
  • LED light rated up to 50,000 hours life, illumination equal to a 20 watt tungsten bulb
  • Separate low position coarse and fine focusing controls
  • 4-hole objective turret, reverse position, ball-bearing mounted for smooth, precise changes in magnification
A new standard of precision optics, highly engineered mechanics and very rugged construction, all at a little more cost than a budget microscope. Objective turret (nosepiece) is in reverse position to permit easier changing and positioning of specimen slides. Preferred by teachers, the rechargeable LED illumination system provides optimum image brightness, virtually no bulb heat and has a 10,000-hour rated bulb life.