UVBioTAG™ Swab, Microbiologics
Supplier: Microbiologics
UVBioTAG™
01224UV-S 01227UV-S 01226UV-S 01223UV-S
75831-376KT 445.85 USD
75831-376 75831-372 75831-374 75831-370
UVBioTAG™ Swab, Microbiologics
Controls and Calibration Verification Materials
Applications include Enumeration Methods, Detection Methods, Demonstration of Capability (DOC), Media QC, and Validation/Verification.
  • Cultures visibly fluoresce under ultraviolet (UV) light to differentiate QC strains from true contaminants
  • Strains are tested using proteomic, phenotypic, and functional methods to ensure their authenticity, stability, and compatibility with common detection technologies
  • GFP reporter is integrated into the chromosome to provide superior stability
  • Instant dissolve pellet reduces preparation time
  • Easy and economical refrigerated storage
  • Online Certificate of Analysis provides detailed strain information
  • Traceability to reference cultures ensures authenticity
UV-BioTag™ microorganisms contain a Green Fluorescent Protein (GFP) marker that produces fluorescence of the culture visible under UV light. This allows for easy differentiation of these control strains from contaminants. UV-BioTAG™ Swab kits include six swabs each containing one lyophilized pellet of an individual microorganism strain.
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the website. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.