Equine Serum, Equitech- Bio
Supplier: Equitech-Bio
SE30-0100 SE30-0500 ES05-0100 ES05-0500
103219-550EA 88.85 USD
103219-550 103219-554 103219-556 103219-558 103219-560 103219-562
Equine Serum, Equitech- Bio
Sera (Serum)
All serum is extensively scrutinized, including negative EIA as release criteria
  • Diet and worming are controlled and documented to ensure a healthy animal
  • Sterility tested
  • Endotoxin and mycoplasma testing available
  • Lot to lot consistency
Raw serum is non-sterile. Low hemoglobin levels. Physiochemical constituents measured to ensure lot-to-lot consistency. Serum from specific breeds are available by request. Sterile-Filtered serum is sterility tested and sterile-filtered to 0.2 micron absolute. Endotoxin and mycoplasma testing available. Heat-inactivation is available if desired.
All horses are of U.S. origin.