2-Propanol 99% antiseptic, cleanser, Hydrox Laboratories

Supplier: Healthcare Products
Danger

Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol

HDX A0053 HDXD0052
75830-308CS 282.12 USD
75830-308 75830-312
2-Propanol 99% antiseptic, cleanser, Hydrox Laboratories
2-Propanol

  • Topical antiseptic
  • Active ingredient: isopropyl alcohol

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Flash Pt: 12
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
ADR: 3 II
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR