1,8-Diazabicyclo[5.4.0]undec-7-ene ≥95%
Supplier: Matrix Scientific
Synonyms:
DBU
, 2,3,4,6,7,8,9,10-Octahydropyrimido[1,2-a]azepine
072922-500G
102833-570EA
230.77
USD
102833-570
1,8-Diazabicyclo[5.4.0]undec-7-ene ≥95%
1,8-Diazabicyclo[5.4.0]undec-7-ene
TCA listed: T
Hazard: Irritant
Formula:
C₉H₁₆N₂ MW: 152.24 g/mol Boiling Pt: 261 °C Density: 1.02 Flash Pt: 116 °C |
MDL Number:
MFCD00006930 CAS Number: 6674-22-2 UN: 3267 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...