4-Ethyl-2-(1H-pyrrol-1-yl)-1,3-thiazole-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

115180-500MG 115180-1G
102895-530EA 693.85 USD
102895-530 102895-532
4-Ethyl-2-(1H-pyrrol-1-yl)-1,3-thiazole-5-carboxylic acid ≥95%
4-Ethyl-2-(1H-pyrrol-1-yl)-1,3-thiazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂S
MW: 222.27 g/mol
MDL Number: MFCD26130172
CAS Number: 1610377-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR