1-[6-(2-Fluorophenyl)pyridazin-3-yl]pyrrolidin-3-amine ≥95%

Supplier: Matrix Scientific

115137-500MG 115137-1G
102891-778EA 693.85 USD
102891-778 102895-474
1-[6-(2-Fluorophenyl)pyridazin-3-yl]pyrrolidin-3-amine ≥95%
1-[6-(2-Fluorophenyl)pyridazin-3-yl]pyrrolidin-3-amine

Hazard: Irritant

Formula: C₁₄H₁₅FN₄
MW: 258.3 g/mol
MDL Number: MFCD28023531
CAS Number: 1610377-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR