1-{[1-(2-Methoxyphenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride ≥95%

Supplier: Matrix Scientific

115125-500MG 115125-1G
102899-078EA 693.85 USD
102899-078 102899-080
1-{[1-(2-Methoxyphenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride ≥95%
1-{[1-(2-Methoxyphenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂N₅O
MW: 346.26 g/mol
MDL Number: MFCD28023523
CAS Number: 1610377-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR