Isoquinoline-1-carbonyl chloride ≥97%
Supplier: Matrix Scientific
113769-1G
102898-068EA
813.85
USD
102898-068
Isoquinoline-1-carbonyl chloride ≥97%
Isoquinoline-1-carbonyl chloride
Hazard: Irritant
Formula:
C₁₀H₆ClNO |
MDL Number:
MFCD16077292 CAS Number: 180411-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...