10-Phenyl-3,4,9,10-tetrahydrobenzo[b]cyclopenta[e][1,4]diazepin-1(2H)-one

Supplier: Matrix Scientific

080623-500MG 080623-1G 080623-5G
102670-442EA 193.85 USD
102670-442 102670-444 102670-446
10-Phenyl-3,4,9,10-tetrahydrobenzo[b]cyclopenta[e][1,4]diazepin-1(2H)-one
10-Phenyl-3,4,9,10-tetrahydrobenzo[b]cyclopenta[e][1,4]diazepin-1(2H)-one

Hazard: Irritant

Formula: C₁₈H₁₆N₂O
MDL Number: MFCD22374927
CAS Number: 1374509-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR