2-Methyl-3-oxo-3,5,6,7-tetrahydro-2H-cyclopenta[c]pyridine-4-carbonitrile
Supplier: Matrix Scientific
080588-500MG
080588-1G
102670-288EA
387.69
USD
102670-288
102670-290
2-Methyl-3-oxo-3,5,6,7-tetrahydro-2H-cyclopenta[c]pyridine-4-carbonitrile
2-Methyl-3-oxo-3,5,6,7-tetrahydro-2H-cyclopenta[c]pyridine-4-carbonitrile
Hazard: Irritant
Formula:
C₁₀H₁₀N₂O |
MDL Number:
MFCD22374914 CAS Number: 1374509-63-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...