Tegaderm™ CHG Chlorhexidine Gluconate I.V. Securement Dressing, 3M™
Supplier: Healthcare Products
MMM 1660 MMM 1657 MMM 1658 MMM 1659
75786-812CS 2618.07 USD
75786-812 75786-814 75786-816 75786-818
Tegaderm™ CHG Chlorhexidine Gluconate I.V. Securement Dressing, 3M™
First Aid Supplies Wound Dressings
Dressings combine antimicrobial protection, securement, site visibility, and breathability into a single product.
  • Transparent film and gel pad allows continuous site observation
  • Promotes moisture evaporation and enhances securement
  • CHG gel pad provides immediate and continuous protection