H-800 Series Full Brim Hard Hats, 3M™
Supplier: 3M
7000144964 7000144975 7100162736 7100162737 7010296269 7100162734 7100162731 7000144985 7000144996 7100162732 7100162733 7100162735
10756-270CS 752.56 USD
10756-270 10756-228 10756-238 10756-226 10756-268 10756-224 10756-234 10756-266 10756-232 10756-264 10756-230 10756-262
H-800 Series Full Brim Hard Hats, 3M™
Head Protection Safety Helmets
The H-800 Series Full Brim Hard Hat with Ratchet System is comfortable, tough, and lightweight protective headgear.
  • Protective headgear helps protect the head from small falling objects and fixed objects that may be bumped into
  • 4-point suspension system has unique features that allows for a customized height and fit that best suits the individual needs of the wearer
Accessories information: Easy-opening clips allow goggle attachment to virtually any hard hat. Clips help keep goggles from being lost, scratched, or misplaced and allows workers to place goggles below the brim when in use, or slide above the brim when not in use. Plastic clips mount quickly with contact adhesive.