Sodium hydrogen carbonate concentrate 0.5M IC

Supplier: INORGANIC VENTURES MS

Synonyms: Sodium bicarbonate

BICARB-100ML BICARB-500ML
101372-426EA 46.74 USD
101372-426 101372-428
Sodium hydrogen carbonate concentrate 0.5M IC
Sodium hydrogen carbonate

0.5M Sodium Bicarbonate Concentrate for IC Eluent Preparation


  • Appearance: Clear
  • Matrix: Water


Delivery information: One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).

Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270
MDL Number: MFCD00003528
CAS Number: 144-55-8
Merck Index: 13,08655

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR