Chromium ICP Standard, Inorganic Ventures
Supplier: Inorganic Ventures
CGCR(3)10-500ML
101446-574EA 236.45 USD
101446-574
Chromium ICP Standard, Inorganic Ventures
Standards Cationic Standards Chromium Standards
Certified reference material for Chromium, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).