Methyl ethyl ketone ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
Synonyms:
2-Butanone
BDH25643.400
BDH25643.400CS
1468.54
USD
BDH25643.400
Methyl ethyl ketone ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Methyl ethyl ketone
Formula:
C₂H₅COCH₃ MW: 72.11 g/mol Boiling Pt: 79.6 °C (1013 hPa) Melting Pt: –86 °C Flash Pt: –4 °C |
MDL Number:
MFCD00011648 CAS Number: 78-93-3 UN: 1193 ADR: 3,II |
Specification Test Results
Assay (by GC-FID) | Min 99.5% |
Color | Max 10 APHA |
Residue after evaporation | Max 5 ppm |
Trace organic impurity sec-Butyl Alcohol | Max 0.2% |
Titrable acid | Max 0.0005 meq/g |
Water (by KF coulometry) | Max 0.8% |
Transmittance at 329 nm | Min 10 T(%) |
Transmittance at 335 nm | Min 30 T(%) |
Transmittance at 340 nm | Min 80 T(%) |
Transmittance at 344 nm | Min 90 T(%) |
Transmittance at 350 nm | Min 98 T(%) |
Transmittance at 375 nm | Min 98 T(%) |
Transmittance at 400 nm | Min 98 T(%) |
Optical absorbance at 329 nm | Max 1.00 AU |
Optical absorbance at 335 nm | Max 0.50 AU |
Optical absorbance at 340 nm | Max 0.10 AU |
Optical absorbance at 344 nm | Max 0.05 AU |
Optical absorbance at 350 nm | Max 0.01 AU |
Optical absorbance at 375 nm | Max 0.01 AU |
Optical absorbance at 400 nm | Max 0.01 AU |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...