FUTURO™ Stirrup Ankle Brace, 3M™, National Distribution and Contracting
Supplier: National Distribution and Contracting
FUTURO™
MMM48442EN
75786-674CS 528.87 USD
75786-674
FUTURO™ Stirrup Ankle Brace, 3M™, National Distribution and Contracting
Ergonomic Supports Ankle Supports
Provides comfortable, rigid support and protection in case of a moderate-to-severe injury, for weak ankles and/or for post-cast treatment.
  • Anatomically designed shell helps limit lateral motion post-cast or after an acute injury
  • Inflatable air cells help enhance circulation and reduce swelling
  • Use on left or right ankle