Reston™ Self-Adhering Foam Products, 3M™
Supplier: Healthcare Products
MMM 1561H MMM 1563L MMM 1560M
75786-502CS 340.34 USD
75786-502 75786-504 75786-506
Reston™ Self-Adhering Foam Products, 3M™
First Aid Supplies First Aid Kit Components
Self-adhering open cell foam is designed to protect against skin damage from splints, casts, and prostheses.
  • Versatile padding adheres well to a variety of surfaces
  • Lightweight and conformable padding can be cut, layered, and shaped
Light Support pads are supplied on a roll, and are 0.47 cm (0.1875") thick. Medium Support pads are 1.1 cm (0.4375") thick. High support pads are 2.4 cm (0.9375") thick.
Product is not intended to be used as a pressure reducing device. It can also be used as a padding for beds and chairs