Heat Sealer, Crosstex
Supplier: Crosstex
MHS-002
101101-908EA 2999 USD
101101-908
Heat Sealer, Crosstex
Heat Sealers Bag Sealers
Heat sealers ideal for sealing paper/poly and Tyvek® packaging products, as well as dust covers.
  • Compact and easy to operate
  • Short warm up period
  • Makes 1/2" seal in two seconds
  • Temperature setting can be changed according to sealing products
Delivery information: Supplied with instructions for use.