2-Propanol ≥99.8%

Supplier: KMG

Synonyms: Isopropyl alcoholIsopropyl Rubbing Alcoholsec-PropanolIsopropyl rubbing alcohol2-Hydroxy propanesec-Propyl AlcoholDimethyl carbinol2-HydroxypropanePropane, 2-hydroxy-Propan-2-olIPAIsopropanol

200445 200446
71009-372CS 535.39 USD
71009-372 89400-822
2-Propanol ≥99.8%
2-Propanol

IPA is used to rinse silicon wafers and to leave a clean dry residue free surface usually after organic stripping solutions.

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg

Order Now

Specification Test Results

Assay (IPA) 99.8%
Color 5
Acidity 1 µeq/g
Water 200 ppm
Residue After Evaporation 1000 ppb
Chloride 50 ppb
Phosphate 50 ppb
Aluminum 1 ppb
Antimony 1 ppb
Arsenic 1 ppb
Barium 1 ppb
Beryllium 1 ppb
Bismuth 1 ppb
Boron 1 ppb
Cadmium 1 ppb
Calcium 1 ppb
Chromium 1 ppb
Cobalt 1 ppb
Copper 1 ppb
Gallium 1 ppb
Germanium 1 ppb
Gold 1 ppb
Iron 1 ppb
Lead 1 ppb
Lithium 1 ppb
Magnesium 1 ppb
Manganese 1 ppb
Molybdenum 1 ppb
Nickel 1 ppb
Niobium 1 ppb
Potassium 1 ppb
Silver 1 ppb
Sodium 1 ppb
Strontium 1 ppb
Tantalum 1 ppb
Thallium 1 ppb
Tin 1 ppb
Titanium 1 ppb
Vanadium 1 ppb
Zinc 1 ppb
Zirconium 1 ppb
0.20µ Particle Count 350 par/ml
0.50µ Particle Count 17 par/ml
1.00µ Particle Count 5 par/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR