High Recovery Vials, Borosilicate, Chemglass
Supplier: Chemglass
CG-4916-35 CG-4916-31 CG-4916-30 CG-4916-32 CG-4916-02 CG-4916-06 CG-4916-34 CG-4916-03 CG-4916-05 CG-4916-33 CG-4916-04 CG-4916-01
89166-728PK 574.95 USD
89166-728 30622-688 89166-718 30622-698 30622-692 89166-720 30622-690 89166-722 30622-696 89166-724 89166-726 30622-694
High Recovery Vials, Borosilicate, Chemglass
Vials
High recovery vial with a conical bottom for complete sample recovery.
  • Boxes are marked with alpha-numeric coordinates for fast and easy vial identification
  • Available complete with open-top caps or vials only
These vials are the same size or slightly smaller on the OD to allow them to be interchanged with any racks or blocks that accept standard dram or scintillation vials. Packed in an SBS coated white tray complete with SBS partition.