Gensolve™ 475 for cleaning
Supplier: KMG
200487
89400-780CS
2104.59
USD
89400-780
Gensolve™ 475 for cleaning
GenSolve™
The Gensolve product line are organic stripping compositions that remove hydrophillic polymeric organic coating, such as photoresists and their residues from inorganic substrates.
Specification Test Results
Assay (H2O) | 1,00% |
Color | 250 |
Refractive Index | 1.40 - 1.50 |
Specific Gravity | 1.02 - 1.10 |
Surface Tension | 43.0 - 52.0 dynes/cm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...