Nickel Single Element Standards for ICP and ICP-MS, Ultra Scientific
Supplier: AGILENT TECHNOLOGIES
ICP-028 ICP-128
UL-ICP-028EA 89.28 USD
UL-ICP-028 UL-ICP-128
Nickel Single Element Standards for ICP and ICP-MS, Ultra Scientific
Standards Cationic Standards Nickel Standards
The ICP standard is manufactured and verified under Ultra’s ISO 9001 registered quality system. It is prepared from starting materials having a purity of 99.999% (where possible), high purity acids, and ASTM Type I water (18 megaohm).
It is traceable to NIST standard reference materials (SRM) whenever possible.
Documentation data includes elemental identity, trace metal impurities, and final element concentration by ICP; traceability data to NIST SRM by both wet chemical assay and ICP on final solution lot; and final solution density.