Disposable Blade Carriers, Thermo Scientific
Supplier: Thermo Scientific
705800 705050 705100 705100 705050
16002-424EA 4883.98 USD
16002-424 10755-210 CA16002-424 CA84000-446 84000-446
Disposable Blade Carriers, Thermo Scientific
Razors and Blades
Utilize low-and high-profile blades with the quick clamp system of Thermo Scientific™ Disposable Blade Carriers
  • Quick clamp system for high-and low-profile blades
  • Adjustable clearance angle
  • Moveable knife guard covers the blade entirely in any position
  • Knife carrier base included
Updated models (long stroke and version two bar guideways) include removable angle adjustment and blade clamp levers. Blade angle and blade adjustment levers are removable to help protect user settings (Version two and three units only). Concentric lever locks onto the dovetail to help reduce accidental movement of the holder (Version two and three units only).