Ward's® Live Azotobacter vinelandii Culture
Supplier: VWR
WARD470179-050
470179-050EA 12.95 USD
470179-050
Ward's® Live Azotobacter vinelandii Culture
Specimens Live Specimens Bacteria - Living Material
Azotobacter vinelandii is an aerobic organism normally found in soil, and is commonly known for its ability to fix nitrogen, converting it to ammonia.
  • Biosafety Level: 1
  • Gram Stain: Negative
  • Morphology: Round rods, many pairs
  • Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: "Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout