2-Azido-N-(3-chlorophenyl)acetamide ≥95%
Supplier: Matrix Scientific
115162-500MG
115162-1G
102892-178EA
693.85
USD
102892-178
102892-180
2-Azido-N-(3-chlorophenyl)acetamide ≥95%
2-Azido-N-(3-chlorophenyl)acetamide
Hazard: Irritant
Formula:
C₈H₇ClN₄O |
MDL Number:
MFCD14652257 CAS Number: 215320-57-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...