2-(3-Nitrophenyl)propanoic acid ≥97%
Supplier: Matrix Scientific
114443-1G
102894-050EA
1298.46
USD
102894-050
2-(3-Nitrophenyl)propanoic acid ≥97%
2-(3-Nitrophenyl)propanoic acid
Hazard: Irritant
Formula:
C₉H₉NO₄ MW: 195.18 g/mol |
MDL Number:
MFCD11036895 CAS Number: 21762-10-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...