tert-Butyl-3-hydroxy-4-phenylpiperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Boc-3-Hydroxy-4-phenylpiperidine
115464-500MG
115464-1G
115464-5G
102896-350EA
1512.31
USD
102896-350
102896-352
102896-354
tert-Butyl-3-hydroxy-4-phenylpiperidine-1-carboxylate ≥95%
tert-Butyl-3-hydroxy-4-phenylpiperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₆H₂₃NO₃ MW: 277.37 g/mol |
MDL Number:
MFCD08461249 CAS Number: 1000931-04-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...