Diphenyl(trifluoromethyl)sulfonium trifluoromethanesulfonate ≥95%

Supplier: Matrix Scientific

115446-1G 115446-5G
102896-324EA 769.23 USD
102896-324 102896-326
Diphenyl(trifluoromethyl)sulfonium trifluoromethanesulfonate ≥95%
Diphenyl(trifluoromethyl)sulfonium trifluoromethanesulfonate

Hazard: Irritant

Formula: C₁₄H₁₀F₆O₃S₂
MW: 404.35 g/mol
MDL Number: MFCD12912173
CAS Number: 147531-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR