tert-Butyl-2,7-diazaspiro[4.5]decane-2-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 2,7-diazaspiro[4.5]decane-2-carboxylate
115323-250MG
115323-500MG
115323-1G
102899-114EA
769.23
USD
102899-114
102899-116
102899-118
tert-Butyl-2,7-diazaspiro[4.5]decane-2-carboxylate ≥95%
tert-Butyl-2,7-diazaspiro[4.5]decane-2-carboxylate
Hazard: Irritant
Formula:
C₁₃H₂₄N₂O₂ MW: 240.35 g/mol |
MDL Number:
MFCD08234437 CAS Number: 885268-42-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...