(R)-2-(1-Naphthylmethyl)succinic acid-1-methyl ester ≥95%

Supplier: Matrix Scientific

115251-500MG 115251-1G
102899-168EA 590.77 USD
102899-168 102899-170
(R)-2-(1-Naphthylmethyl)succinic acid-1-methyl ester ≥95%
(R)-2-(1-Naphthylmethyl)succinic acid-1-methyl ester

Hazard: Irritant

Formula: C₁₆H₁₆O₄
MW: 272.3 g/mol
MDL Number: MFCD06798981
CAS Number: 119807-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR