6,7-Dimethoxy-2-oxo-1,2-dihydroquinoline-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

115167-500MG 115167-1G
102892-198EA 693.85 USD
102892-198 102895-498
6,7-Dimethoxy-2-oxo-1,2-dihydroquinoline-3-carbaldehyde ≥95%
6,7-Dimethoxy-2-oxo-1,2-dihydroquinoline-3-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₁₁NO₄
MW: 233.23 g/mol
MDL Number: MFCD07506517
CAS Number: 101382-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR