3-[3-(3-Thienyl)-1,2,4-oxadiazol-5-yl]pyridin-2(1H)-one ≥95%

Supplier: Matrix Scientific

115128-500MG 115128-1G
102895-460EA 693.85 USD
102895-460 102895-462
3-[3-(3-Thienyl)-1,2,4-oxadiazol-5-yl]pyridin-2(1H)-one ≥95%
3-[3-(3-Thienyl)-1,2,4-oxadiazol-5-yl]pyridin-2(1H)-one

Hazard: Irritant

Formula: C₁₁H₇N₃O₂S
MW: 245.26 g/mol
MDL Number: MFCD22391001
CAS Number: 1389455-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR