4-(2,4-Dioxo-1,4-dihydrothieno[3,2-d]pyrimidin-3(2H)-yl)butanoic acid ≥97%

Supplier: Matrix Scientific

115113-500MG 115113-1G
102895-444EA 693.85 USD
102895-444 102895-446
4-(2,4-Dioxo-1,4-dihydrothieno[3,2-d]pyrimidin-3(2H)-yl)butanoic acid ≥97%
4-(2,4-Dioxo-1,4-dihydrothieno[3,2-d]pyrimidin-3(2H)-yl)butanoic acid

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₄S
MW: 254.27 g/mol
MDL Number: MFCD04063015
CAS Number: 702664-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR