4,5-Dimethylsalicylaldehyde ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Hydroxy-4,5-dimethylbenzaldehyde
113563-1G
102892-442EA
813.85
USD
102892-442
4,5-Dimethylsalicylaldehyde ≥97%
4,5-Dimethylsalicylaldehyde
Hazard: Irritant
Formula:
C₉H₁₀O₂ MW: 150.175 g/mol |
MDL Number:
MFCD06740693 CAS Number: 1666-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...