1-(2-Diethylaminoethyl)piperazine ≥97%

Supplier: Matrix Scientific

113536-1G 113536-10G
102892-380EA 60 USD
102892-380 102892-382
1-(2-Diethylaminoethyl)piperazine ≥97%
1-(2-Diethylaminoethyl)piperazine

Hazard: Irritant

Formula: C₁₀H₂₃N₃
MDL Number: MFCD00167968
CAS Number: 4038-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR