4-Propionylbenzoic acid ≥97%
Supplier: Matrix Scientific
087027-1G
087027-2G
087027-5G
102852-058EA
490.77
USD
102852-058
102852-060
102852-062
4-Propionylbenzoic acid ≥97%
4-Propionylbenzoic acid
Hazard: Irritant
Formula:
C₁₀H₁₀O₃ |
MDL Number:
MFCD02260861 CAS Number: 4219-55-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...