2',3'-Dimethyl-2,2,2-trifluoroacetophenone ≥97%
Supplier: Matrix Scientific
Synonyms:
1-(2,3-Dimethylphenyl)-2,2,2-trifluoroethanone
086905-5G
086905-1G
086905-2G
102851-332EA
347.69
USD
102851-332
102851-336
102851-334
2',3'-Dimethyl-2,2,2-trifluoroacetophenone ≥97%
2',3'-Dimethyl-2,2,2-trifluoroacetophenone
Hazard: Refrigerate
Formula:
C₁₀H₉F₃O MW: 202.18 g/mol |
MDL Number:
MFCD01319983 CAS Number: 886369-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...