Methyl-2-(6-methoxynicotinoyl)benzoate ≥97%

Supplier: Matrix Scientific

086637-1G 086637-2G 086637-5G
102849-736EA 795.38 USD
102849-736 102849-738 102849-740
Methyl-2-(6-methoxynicotinoyl)benzoate ≥97%
Methyl-2-(6-methoxynicotinoyl)benzoate

Hazard: Irritant

Formula: C₁₅H₁₃NO₄
MDL Number: MFCD07699472
CAS Number: 898786-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR