2,5-Dibromo-3-decylthiophene ≥97%
Supplier: Matrix Scientific
086502-25G
086502-50G
102848-930EA
5233.85
USD
102848-930
102848-932
2,5-Dibromo-3-decylthiophene ≥97%
2,5-Dibromo-3-decylthiophene
Hazard: Irritant
Formula:
C₁₄H₂₂Br₂S |
MDL Number:
MFCD00274314 CAS Number: 158956-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...