Ethyl-5-methylthiophene-2-glyoxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-(5-methylthiophen-2-yl)-2-oxoacetate
086421-2G
102848-588EA
827.69
USD
102848-588
Ethyl-5-methylthiophene-2-glyoxylate ≥97%
Ethyl-5-methylthiophene-2-glyoxylate
Hazard: Irritant
Formula:
C₉H₁₀O₃S MW: 198.24 g/mol |
MDL Number:
MFCD07782947 CAS Number: 50845-87-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...