5-(Trifluoromethyl)anthranilic acid ≥95%

Supplier: Matrix Scientific

086154-1G 086154-5G
102847-084EA 38.46 USD
102847-084 102847-086
5-(Trifluoromethyl)anthranilic acid ≥95%
5-(Trifluoromethyl)anthranilic acid

Hazard: Irritant

Formula: C₈H₆F₃NO₂
Melting Pt: 191…192 °C
MDL Number: MFCD01569537
CAS Number: 83265-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR