Dimethyl sulfoxide-[D₆] ≥99%
Supplier: Matrix Scientific
Synonyms:
Dimethyl sulfoxide-[²H₆]
, Hexadeuterodimethyl sulfoxide
, DMSO-D6
, DMSO-d6
, Dimethyl sulphoxide-d6
, Dimethyl sulfoxide-d6
, Dimethyl sulphoxide-[²H₆]
, Hexadeuterodimethyl sulphoxide
086137-50G
086137-10G
102847-070EA
607.69
USD
102847-070
102847-068
Dimethyl sulfoxide-[D₆] ≥99%
Dimethyl sulphoxide-[D6]
Hazard: Irritant
Formula:
D₃CS(O)CD₃ MW: 84.09 g/mol Boiling Pt: 189 °C Melting Pt: 18.4 °C Density: 1.19 Flash Pt: 87 °C |
MDL Number:
MFCD00002090 CAS Number: 2206-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...